3,5-二甲氧基苯硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
Synonyms:
3,5-Dimethoxybenzeneboronic acid
D3513-1G
D3513-25G
D3513-5G
TCIAD3513-1GEA
0
CNY
TCIAD3513-1G
TCIAD3513-25G
TCIAD3513-5G
3,5-二甲氧基苯硼酸 (contains varying amounts of Anhydride)
3,5-二甲氧基苯硼酸
分子式:
C₈H₁₁BO₄ 分子量: 181.98 g/mol 溶点: 202°C |
MDL编号:
MFCD03095127 CAS号: 192182-54-0 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...