(E)-苯甲醛肟 ≥90.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(E)-Benzaldehyde oxime, alpha-Benzaldoxime
B0011-25G
B0011-500G
TCIAB0011-25GEA
0
CNY
TCIAB0011-25G
TCIAB0011-500G
(E)-苯甲醛肟 ≥90.0% (by GC)
(E)-苯甲醛肟
分子式:
C₇H₇NO 分子量: 121.14 g/mol 沸点: 123°C 存储条件: Refrigerator |
MDL编号:
MFCD00002119 CAS号: 622-31-1 EINECS: 213-261-2 |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...