二溴氯甲烷 ≥95.0% (by GC) stabilised

供应商: TCI
Warning

Synonyms: 一氯二溴甲烷

Stabilization: Stabilised with ethanol

C0854-25G C0854-5G
TCIAC0854-25GEA 0 CNY
TCIAC0854-25G TCIAC0854-5G
二溴氯甲烷 ≥95.0% (by GC) stabilised
二溴氯甲烷
分子式: Br₂ClCH
沸点: 122°C
溶点: –22 °C
密度: 2,445 g/cm³ (15 °C)
存储条件: Refrigerator
MDL编号: MFCD00000820
CAS号: 124-48-1
EINECS: 204-704-0
UN号: 3077
ADR: 9,III
Merck Index: 14,02135
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99312119282f4f5e418705bff82a1f2a1702d9b7&vid=aaa9eef671fb0aca2faa0ac2a7ff471ada715347&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR