尿嘧啶 ≥98.0% (by HPLC, titration analysis)

供应商: TCI

Synonyms: 2,4-二羟基嘧啶, 2,4(1H,3H)-嘧啶二酮, 灭胞素

U0013-25G U0013-500G
TCIAU0013-25GEA 0 CNY
TCIAU0013-25G TCIAU0013-500G
尿嘧啶 ≥98.0% (by HPLC, titration analysis)
尿嘧啶
分子式: C₄H₄N₂O₂
沸点: 379 °C (1013 hPa)
溶点: 194 °C
密度: 0,99744 g/cm³ (25 °C)
存储条件: Ambient
MDL编号: MFCD00006016
CAS号: 66-22-8
EINECS: 200-621-9
Merck Index: 14,09850
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0205dd888f415bc44387cd510365e2633d90ae07&vid=0205dd888f415bc44387cd510365e2633d90ae07&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Sodium hydroxide solution almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR