3-溴丙胺 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3-氨基丙基溴

B1263-25G B1263-500G
TCIAB1263-25GEA 0 CNY
TCIAB1263-25G TCIAB1263-500G
3-溴丙胺 ≥98.0% (by titrimetric analysis)
3-溴丙胺
分子式: BrH₂CCH₂CH₂NH₃Br
分子量: 218.92 g/mol
MDL编号: MFCD00012912
CAS号: 5003-71-4
EINECS: 225-675-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aab9bb0f7da6b6725d00a9a7afd7c83aff8b0627&vid=aab9bb0f7da6b6725d00a9a7afd7c83aff8b0627&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Brown powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR