1,3,5-三(2-羟乙基)异氰尿酸酯 ≥98.0% (以总氮计)
供应商: TCI
|
Warning
|
T0674-25G
T0674-500G
TCIAT0674-25GEA
0
CNY
TCIAT0674-25G
TCIAT0674-500G
1,3,5-三(2-羟乙基)异氰尿酸酯 ≥98.0% (以总氮计)
1,3,5-三(2-羟乙基)异氰尿酸酯
分子式:
C₉H₁₅N₃O₆ 沸点: 527 °C (1013 hPa) 溶点: 133…136 °C |
MDL编号:
MFCD00003549 CAS号: 839-90-7 EINECS: 212-660-9 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 134.0 to 138.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...