乙酰丙酮钴(III) ≥98.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: 乙酰丙酮合钴, 三(2,4-戊二酮)钴, Co(acac)3, 三乙酰丙酮钴

C0374-25G C0374-500G
TCIAC0374-25GEA 0 CNY
TCIAC0374-25G TCIAC0374-500G
乙酰丙酮钴(III) ≥98.0% (by titrimetric analysis)
乙酰丙酮钴(III)
分子式: C₁₅H₂₁CoO₆
分子量: 356.26 g/mol
MDL编号: MFCD00013488
CAS号: 21679-46-9
EINECS: 244-527-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45f6c2c23cf0b4c8c2361be2d369e0127bb6ac3e&vid=45f6c2c23cf0b4c8c2361be2d369e0127bb6ac3e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Green to Dark green powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR