富勒烯-C60 ≥99.5% (by HPLC), pure
供应商: TCI
Synonyms:
巴克明斯特·富勒烯
B1641-1G
B1641-100MG
TCIAB1641-1GEA
0
CNY
TCIAB1641-1G
TCIAB1641-100MG
富勒烯-C60 ≥99.5% (by HPLC), pure
富勒烯-C60
分子式:
C₆₀ 分子量: 720.66 g/mol |
MDL编号:
MFCD00151408 CAS号: 99685-96-8 Merck Index: 14,01462 |
|
产品技术规格
Appearance | Black powder to crystal |
Purity (HPLC) | min. 99.5 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...