乙醇钛 (contains 35% Tetraisopropyl Orthotitanate at maximum)

供应商: TCI
Warning

Synonyms: 钛酸乙酯, 原钛酸四乙酯

T0967-25ML T0967-500ML
TCIAT0967-25MLEA 0 CNY
TCIAT0967-25ML TCIAT0967-500ML
乙醇钛 (contains 35% Tetraisopropyl Orthotitanate at maximum)
乙醇钛
分子式: C₈H₂₀O₄Ti
分子量: 228.12 g/mol
沸点: 236°C
闪点: 33°C
MDL编号: MFCD00009071
CAS号: 3087-36-3
EINECS: 221-410-8
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=faffe82b73fcdbf04cf64cc7bf692bd5da767c57&vid=faffe82b73fcdbf04cf64cc7bf692bd5da767c57&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Ignition residue 33.0 to 36.0 %
Assay of Tetraisopropyl Orthotitanate 15.0 to 35.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR