辛酸乙酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
亚羊脂酸乙酯
O0030-25ML
O0030-500ML
TCIAO0030-25MLEA
0
CNY
TCIAO0030-25ML
TCIAO0030-500ML
辛酸乙酯 ≥98.0% (by GC)
辛酸乙酯
分子式:
C₁₀H₂₀O₂ 沸点: 208°C 溶点: -43°C 密度: 0,87 g/cm³ (20 °C) 闪点: 92°C 存储条件: Ambient |
MDL编号:
MFCD00009552 CAS号: 106-32-1 EINECS: 203-385-5 Merck Index: 14,03778 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...