4-羟基苯甲酸丁酯 ≥99.0% (by GC, titration analysis) for biochemical research
供应商: TCI
|
Warning
|
Synonyms:
对羟基苯甲酸正丁酯, 对羟基苯甲酸丁酯
B3771-25G
B3771-5G
TCIAB3771-25GEA
0
CNY
TCIAB3771-25G
TCIAB3771-5G
4-羟基苯甲酸丁酯 ≥99.0% (by GC, titration analysis) for biochemical research
4-羟基苯甲酸丁酯
分子式:
C₁₁H₁₄O₃ 分子量: 194.23 g/mol 沸点: 156°C 溶点: 73 |
MDL编号:
MFCD00016478 CAS号: 94-26-8 EINECS: 202-318-7 Merck Index: 14,01584 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 69.0 to 72.0 deg-C |
Protein impurity test (SDS-PAGE) | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...