4-(三氟甲氧基)苯硼酸 (contains varying amounts of Anhydride)
供应商: TCI
|
Warning
|
T1773-25G
T1773-5G
TCIAT1773-25GEA
0
CNY
TCIAT1773-25G
TCIAT1773-5G
4-(三氟甲氧基)苯硼酸 (contains varying amounts of Anhydride)
4-(三氟甲氧基)苯硼酸
分子式:
C₇H₆BF₃O₃ |
MDL编号:
MFCD01074648 CAS号: 139301-27-2 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...