4-乙基苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: (4-Ethylphenyl)boronic acid, (4-Ethylphenyl)metaboric acid, 4-Ethylbenzeneboronic acid, Dihydroxy(4-ethylphenyl)borane

E0720-25G E0720-5G
TCIAE0720-25GEA 0 CNY
TCIAE0720-25G TCIAE0720-5G
4-乙基苯硼酸 (contains varying amounts of Anhydride)
4-乙基苯硼酸
分子式: C₈H₁₁BO₂
密度: 1,125 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00859377
CAS号: 63139-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a90044ca8c78de716474d63d906cb1c3e3da9fd&vid=9a90044ca8c78de716474d63d906cb1c3e3da9fd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (Neutralization titration) 97.0 to 114.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR