嘧啶 ≥98.0% (通过GC,滴定分析) free base

供应商: TCI
Warning

P0662-1G P0662-5G
TCIAP0662-1GEA 0 CNY
TCIAP0662-1G TCIAP0662-5G
嘧啶 ≥98.0% (通过GC,滴定分析) free base
嘧啶
分子式: C₄H₄N₂
沸点: 123°C
溶点: 21 °C
密度: 1,529 g/cm³ (20 °C)
闪点: 34°C
MDL编号: MFCD00006059
CAS号: 289-95-2
EINECS: 206-026-0
UN号: 1325
ADR: 4.1,III
Merck Index: 14,07987
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5179419db4811f8ea0d5cbbf03bf7acab60bb19&vid=088cd42881db91f8e6579301e1c6762d3438752d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR