Dibutyl sulphate ≥96.0% (by GC) alkylating agent

供应商: TCI
Danger

Synonyms: Di-n-butylsulphate

D1633-25G D1633-500G
TCIAD1633-25GEA 0 CNY
TCIAD1633-25G TCIAD1633-500G
Dibutyl sulphate ≥96.0% (by GC) alkylating agent
Dibutyl sulphate
分子式: C₈H₁₈O₄S
分子量: 210.29 g/mol
沸点: 141°C
存储条件: Ambient
MDL编号: MFCD00059420
CAS号: 625-22-9
UN号: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9c7e7c5eb7f11ed5b2f9fd598cd81ad37e37bb9&vid=252820cd8efed8866804bbd6e72f7ffe7122b99c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR