5-Bromo-8-quinolinol ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: 5-Bromoquinolin-8-ol

B3324-25G B3324-5G
TCIAB3324-25GEA 0 CNY
TCIAB3324-25G TCIAB3324-5G
5-Bromo-8-quinolinol ≥96.0% (by GC)
5-Bromo-8-quinolinol
分子式: C₉H₆BrNO
分子量: 224.06 g/mol
MDL编号: MFCD00457011
CAS号: 1198-14-7
EINECS: 214-829-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f783724f9e0cec859bda1f8f02819997fbced506&vid=f783724f9e0cec859bda1f8f02819997fbced506&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR