蔗糖八乙酸酯 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 八乙酸蔗糖酯

S0052-25G S0052-500G
TCIAS0052-25GEA 0 CNY
TCIAS0052-25G TCIAS0052-500G
蔗糖八乙酸酯 ≥98.0% (by titrimetric analysis)
蔗糖八乙酸酯
分子式: C₂₈H₃₈O₁₉
分子量: 678.6 g/mol
存储条件: Refrigerator
MDL编号: MFCD00006623
CAS号: 126-14-7
EINECS: 204-772-1
Merck Index: 14,08881
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ef8bcfc89738503f5e1a3e59082a2c8af41f513&vid=5ef8bcfc89738503f5e1a3e59082a2c8af41f513&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 96.0 area%
Purity (Neutralization back titration) min. 98.0 %
Melting point 85.0 to 89.0 deg-C
Specific rotation [a]20/D +58.0 to +62.0 deg (C=2, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR