1-Amino-4-naphthol hydrochloride 90%, 技术级
供应商: Thermo Fisher Scientific
|
Warning
|
158510050
158510250
ACRO158510050EA
0
CNY
ACRO158510050
ACRO158510250
1-Amino-4-naphthol hydrochloride 90%, 技术级
1-Amino-4-naphthol hydrochloride
分子式:
C₁₀H₁₀ClNO 溶点: 273 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00012562 CAS号: 5959-56-8 EINECS: 227-727-8 |
产品技术规格
Appearance | Pink to grey Powder |
Infrared spectrum | Conforms |
HPLC | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...