Diethyl (3-methoxybenzyl)phosphonate ≥98.0% (通过GC)

供应商: TCI

D3326-25G D3326-5G
TCIAD3326-25GEA 0 CNY
TCIAD3326-25G TCIAD3326-5G
Diethyl (3-methoxybenzyl)phosphonate ≥98.0% (通过GC)
Diethyl (3-methoxybenzyl)phosphonate
分子式: C₁₂H₁₉O₄P
沸点: 135°C
MDL编号: MFCD08276363
CAS号: 60815-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72631f7f6ca335e1e070386de7dcb2e7099e1da&vid=e72631f7f6ca335e1e070386de7dcb2e7099e1da&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR