Diethyl (4-iodobenzyl)phosphonate ≥98.0% (by GC)

供应商: TCI

Synonyms: Diethyl 4-iodobenzylphosphonate, (4-Iodobenzyl)phosphonic acid diethyl ester

D3689-25G D3689-5G
TCIAD3689-25GEA 0 CNY
TCIAD3689-25G TCIAD3689-5G
Diethyl (4-iodobenzyl)phosphonate ≥98.0% (by GC)
Diethyl (4-iodobenzyl)phosphonate
分子式: C₁₁H₁₆IO₃P
分子量: 354.121 g/mol
MDL编号: MFCD09033365
CAS号: 173443-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd865cb8feb4786a324ab31a87b19d1fb4f3d824&vid=bd865cb8feb4786a324ab31a87b19d1fb4f3d824&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR