4-氯苄基膦酸二乙酯 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Diethyl 4-chlorobenzylphosphonate
D3335-25G
D3335-5G
TCIAD3335-25GEA
0
CNY
TCIAD3335-25G
TCIAD3335-5G
4-氯苄基膦酸二乙酯 ≥97.0% (by GC)
4-氯苄基膦酸二乙酯
分子式:
C₁₁H₁₆ClO₃P 分子量: 262.67 g/mol 沸点: 180°C |
MDL编号:
MFCD00051568 CAS号: 39225-17-7 EINECS: 254-365-8 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...