3-乙酰胺基苯硼酸 (contains varying amounts of Anhydride)

供应商: TCI
Warning

Synonyms: 3-乙酰氨基苯硼酸

A2453-1G A2453-5G
TCIAA2453-1GEA 0 CNY
TCIAA2453-1G TCIAA2453-5G
3-乙酰胺基苯硼酸 (contains varying amounts of Anhydride)
3-乙酰胺基苯硼酸
分子式: C₈H₁₀BNO₃
溶点: 140°C
MDL编号: MFCD00236013
CAS号: 78887-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=172925ed7ba3619345f682302df860442ab25f01&vid=172925ed7ba3619345f682302df860442ab25f01&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR