4-溴代巴豆酸甲酯 85% tech.

供应商: Thermo Fisher Scientific
Danger

Synonyms: Methyl 4-bromobut-2-enoate

126450250 126451000
ACRO126450250EA 0 CNY
ACRO126450250 ACRO126451000
4-溴代巴豆酸甲酯 85% tech.
4-溴代巴豆酸甲酯
分子式: C₅H₇BrO₂
分子量: 179.01 g/mol
沸点: 83…85 °C (13 mmHg)
密度: 1.52 g/cm³
闪点: 91 °C
存储条件: Refrigerator
MDL编号: MFCD00000246
CAS号: 1117-71-1
EINECS: 214-251-0
UN号: 1760
ADR: 8 III

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4970 to 1.5050 (20°C, 589 nm)
GC ≥80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR