2-Amino-4,5-dimethylthiazole hydrobromide ≥98.0% (by HPLC, total nitrogen)
供应商: TCI
|
Warning
|
Synonyms:
2-Amino-4,5-dimethylthiazole Hydrobromide, 4,5-Dimethylthiazol-2-amine hydrobromide
A2062-25G
A2062-5G
TCIAA2062-25GEA
0
CNY
TCIAA2062-25G
TCIAA2062-5G
2-Amino-4,5-dimethylthiazole hydrobromide ≥98.0% (by HPLC, total nitrogen)
2-Amino-4,5-dimethylthiazole hydrobromide
分子式:
C₅H₉BrN₂S |
MDL编号:
MFCD00035212 CAS号: 7170-76-5 EINECS: 230-523-1 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...