2-氧代丙基膦酸二甲酯 ≥95.0% (by GC)
供应商: TCI
Synonyms:
丙酮基磷酸二甲酯
O0208-25G
O0208-5G
TCIAO0208-25GEA
0
CNY
TCIAO0208-25G
TCIAO0208-5G
2-氧代丙基膦酸二甲酯 ≥95.0% (by GC)
2-氧代丙基膦酸二甲酯
分子式:
C₅H₁₁O₄P 分子量: 166.12 g/mol 沸点: 124°C 闪点: 93°C 存储条件: Refrigerator |
MDL编号:
MFCD00008769 CAS号: 4202-14-6 EINECS: 224-110-5 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...