1,2-萘醌-4-磺酸 98%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Folin 试剂, 1,2-萘醌-4-磺酸钠, 3,4-二氢-3,4-二氧代-1-萘磺酸
415400250
415401000
ACRO415400250EA
0
CNY
ACRO415400250
ACRO415401000
1,2-萘醌-4-磺酸 98%
1,2-萘醌-4-磺酸
分子式:
C₁₀H₅NaO₅S 分子量: 260.20 g/mol 溶点: 289 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00001700 CAS号: 521-24-4 EINECS: 208-308-9 |
产品技术规格
Appearance | Orange Powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...