1-萘乙腈 ≥98.0% (通过GC)

供应商: TCI
Danger

N0366-25G N0366-500G
TCIAN0366-25GEA 0 CNY
TCIAN0366-25G TCIAN0366-500G
1-萘乙腈 ≥98.0% (通过GC)
1-萘乙腈
分子式: C₁₂H₉N
MDL编号: MFCD00004041
CAS号: 132-75-2
EINECS: 205-078-1
UN号: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ffb88008216164f1586916b08bafa599d43601&vid=32ffb88008216164f1586916b08bafa599d43601&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR