Diethyl (2-methylbenzyl)phosphonate ≥98.0% (by GC)

供应商: TCI

D3327-25G D3327-5G
TCIAD3327-25GEA 0 CNY
TCIAD3327-25G TCIAD3327-5G
Diethyl (2-methylbenzyl)phosphonate ≥98.0% (by GC)
Diethyl (2-methylbenzyl)phosphonate
分子式: C₁₂H₁₉O₃P
CAS号: 62778-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd654be45a56aa71296193720d62dc31dccd384f&vid=fd654be45a56aa71296193720d62dc31dccd384f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR