膦酰基乙酸甲酯二乙酯 ≥96.0% (by GC)

供应商: TCI
Warning

Synonyms: P,P-膦酰基乙酸甲酯二乙酯

D2873-25G D2873-5G
TCIAD2873-25GEA 0 CNY
TCIAD2873-25G TCIAD2873-5G
膦酰基乙酸甲酯二乙酯 ≥96.0% (by GC)
膦酰基乙酸甲酯二乙酯
分子式: C₇H₁₅O₅P
分子量: 210.17 g/mol
MDL编号: MFCD00009081
CAS号: 1067-74-9
EINECS: 213-938-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a802c04ece94abe50ed7de499a92eaf6b8f0311&vid=7a802c04ece94abe50ed7de499a92eaf6b8f0311&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR