3,4,5-三甲氧基苯乙腈 ≥97.0% (通过GC)
供应商: TCI
|
Danger
|
T1927-25G
T1927-5G
TCIAT1927-25GEA
0
CNY
TCIAT1927-25G
TCIAT1927-5G
3,4,5-三甲氧基苯乙腈 ≥97.0% (通过GC)
分子式:
C₁₁H₁₃NO₃ |
MDL编号:
MFCD00001912 CAS号: 13338-63-1 EINECS: 236-388-5 UN号: 3439 ADR: 6.1,III |
|
产品技术规格
Appearance | White to Light yellow powder to crystaline |
Purity (GC) | min. 97.0 % |
Melting point | 76.0 to 79.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...