Phloretin ≥98.0% (通过HPLC)

供应商: TCI
Warning

P1966-1G P1966-5G
TCIAP1966-1GEA 0 CNY
TCIAP1966-1G TCIAP1966-5G
Phloretin ≥98.0% (通过HPLC)
Phloretin
分子式: C₁₅H₁₄O₅
溶点: 262°C
存储条件: Refrigerator
MDL编号: MFCD00002288
CAS号: 60-82-2
EINECS: 200-488-7
Merck Index: 14,07326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d7df91e4b3b7f11333a7e2148e7b2ab04512002&vid=3d7df91e4b3b7f11333a7e2148e7b2ab04512002&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR