(R)-(-)-3-哌啶甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: (R)-(–)-甲酸哌啶

N0654-1G N0654-5G
TCIAN0654-1GEA 0 CNY
TCIAN0654-1G TCIAN0654-5G
(R)-(-)-3-哌啶甲酸 ≥98.0% (by titrimetric analysis)
(R)-(-)-3-哌啶甲酸
分子式: C₆H₁₁NO₂
分子量: 129.16 g/mol
MDL编号: MFCD01630787
CAS号: 25137-00-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30619bf8c655c4fb71270f3a94f3ede46528afc8&vid=30619bf8c655c4fb71270f3a94f3ede46528afc8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -5.0 to -6.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR