3,4-二甲氧基苯丙酮 ≥97.0% (by GC)

供应商: TCI

Synonyms: 1-(3,4-Dimethoxyphenyl)propan-2-one

V0052-25ML V0052-500ML
TCIAV0052-25MLEA 0 CNY
TCIAV0052-25ML TCIAV0052-500ML
3,4-二甲氧基苯丙酮 ≥97.0% (by GC)
3,4-二甲氧基苯丙酮
分子式: C₁₁H₁₄O₃
分子量: 194.23 g/mol
沸点: 167°C
MDL编号: MFCD00008772
CAS号: 776-99-8
EINECS: 212-285-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d6a76c6c2424dbc1fe50d317b333c0e3ae2795&vid=20d6a76c6c2424dbc1fe50d317b333c0e3ae2795&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR