2-羟乙基二硫化物 ∼50% in water
供应商: TCI
|
Warning
|
Synonyms:
双(2-羟乙基)二硫代, 2,2′-二硫二乙醇
B3587-25G
B3587-500G
TCIAB3587-25GEA
0
CNY
TCIAB3587-25G
TCIAB3587-500G
2-羟乙基二硫化物 ∼50% in water
2-羟乙基二硫化物
分子式:
C₄H₁₀O₂S₂ 分子量: 154.25 g/mol 沸点: 307 溶点: 27.5 |
MDL编号:
MFCD00002906 CAS号: 1892-29-1 EINECS: 217-576-6 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Iodometric titration) | 48.0 to 52.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...