2-氰基苯乙腈 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
α-氰基-邻苯乙腈, 高酞腈
C2247-25G
C2247-5G
TCIAC2247-25GEA
0
CNY
TCIAC2247-25G
TCIAC2247-5G
2-氰基苯乙腈 ≥98.0% (by GC)
2-氰基苯乙腈
分子式:
C₉H₆N₂ 分子量: 142.16 g/mol |
MDL编号:
MFCD00001895 CAS号: 3759-28-2 EINECS: 223-167-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...