羟胺-O-磺酸 ≥90.0% (by titrimetric analysis)

供应商: TCI
Danger

Synonyms: Amino hydrogen sulphate

H0530-25G H0530-500G
TCIAH0530-25GEA 0 CNY
TCIAH0530-25G TCIAH0530-500G
羟胺-O-磺酸 ≥90.0% (by titrimetric analysis)
羟胺-O-磺酸
分子式: H₃NO₄S
分子量: 113.09 g/mol
MDL编号: MFCD00011604
CAS号: 2950-43-8
EINECS: 220-971-6
UN号: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99ae130b7e4c189ebfa8c71393d8fe04fd939524&vid=99ae130b7e4c189ebfa8c71393d8fe04fd939524&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR