4,4′-(六氟异亚丙基)二苯胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2,2-Bis(4-aminophenyl)hexafluoropropane

B1400-25G B1400-5G
TCIAB1400-25GEA 0 CNY
TCIAB1400-25G TCIAB1400-5G
4,4′-(六氟异亚丙基)二苯胺 ≥98.0% (by GC, titration analysis)
4,4′-(六氟异亚丙基)二苯胺
分子式: C₁₅H₁₂F₆N₂
分子量: 334.2600 g/mol
MDL编号: MFCD00039146
CAS号: 1095-78-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0a2c9787b8d4bf7eb785938d1a35c18a6adda86&vid=a0a2c9787b8d4bf7eb785938d1a35c18a6adda86&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 196.0 to 199.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR