1,3-苯二醇单乙酸酯 >50.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-Hydroxyphenyl acetate, Acetic acid 3-Hydroxyphenyl ester

R0009-25G R0009-500G
TCIAR0009-25GEA 0 CNY
TCIAR0009-25G TCIAR0009-500G
1,3-苯二醇单乙酸酯 >50.0% (by GC)
1,3-苯二醇单乙酸酯
分子式: C₈H₈O₃
沸点: 283°C
MDL编号: MFCD00002266
CAS号: 102-29-4
EINECS: 203-022-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfccda0635bdd11c341db8bf79631505756be608&vid=bfccda0635bdd11c341db8bf79631505756be608&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white clear liquid
Purity (GC) min. 50.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR