2,4,6-三溴间苯二酚 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

T2620-1G T2620-5G
TCIAT2620-1GEA 0 CNY
TCIAT2620-1G TCIAT2620-5G
2,4,6-三溴间苯二酚 ≥98.0% (by GC, titration analysis)
2,4,6-三溴间苯二酚
分子式: C₆H₃Br₃O₂
MDL编号: MFCD00009717
CAS号: 2437-49-2
EINECS: 219-442-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b69c3584e849f3dc65e43d23f0482878092f4a&vid=e5b69c3584e849f3dc65e43d23f0482878092f4a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR