(R)-(-)-3-羟基四氢呋喃 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: (R)-Tetrahydrofuran-3-ol

H1332-1G H1332-5G
TCIAH1332-1GEA 0 CNY
TCIAH1332-1G TCIAH1332-5G
(R)-(-)-3-羟基四氢呋喃 ≥98.0% (by GC)
(R)-(-)-3-羟基四氢呋喃
分子式: C₄H₈O₂
沸点: 80°C
闪点: 82°C
MDL编号: MFCD00067101
CAS号: 86087-24-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1896d593030b0f6d429d98cfb0c158ceac7c6a34&vid=1896d593030b0f6d429d98cfb0c158ceac7c6a34&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Specific rotation [a]20/D -17.0 to -20.0 deg (C=2, Methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR