N,O-Dimethylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

D1899-25G D1899-500G
TCIAD1899-25GEA 0 CNY
TCIAD1899-25G TCIAD1899-500G
N,O-Dimethylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)
N,O-Dimethylhydroxylamine hydrochloride
分子式: C₂H₈ClNO
分子量: 97.55 g/mol
MDL编号: MFCD00012485
CAS号: 6638-79-5
EINECS: 229-642-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=141e476ad5f9f075c7121548a8e8e1ac57570c4d&vid=141e476ad5f9f075c7121548a8e8e1ac57570c4d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR