N-甲基羟胺 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: N-Methylhydroxylamine hydrochloride

M0804-25G M0804-5G
TCIAM0804-25GEA 0 CNY
TCIAM0804-25G TCIAM0804-5G
N-甲基羟胺 ≥97.0% (by titrimetric analysis)
N-甲基羟胺
分子式: CH₆ClNO
MDL编号: MFCD00012597
CAS号: 4229-44-1
EINECS: 224-181-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f580eb504886cfe62387e938bbcef2ff03919bcc&vid=f580eb504886cfe62387e938bbcef2ff03919bcc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 83.0 to 88.0 deg-C
Solubility in Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR