2-甲氧基-3-(1-甲基丙基)吡嗪 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-仲丁基-3-甲氧基吡嗪
B2959-1G
B2959-5G
TCIAB2959-1GEA
0
CNY
TCIAB2959-1G
TCIAB2959-5G
2-甲氧基-3-(1-甲基丙基)吡嗪 ≥97.0% (by GC)
2-甲氧基-3-(1-甲基丙基)吡嗪
分子式:
C₉H₁₄N₂O 沸点: 99°C 闪点: 77°C |
MDL编号:
MFCD00006136 CAS号: 24168-70-5 EINECS: 246-050-9 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...