1,4-二氯酞嗪 ≥98.0% (by GC)

供应商: TCI

D3749-1G D3749-5G
TCIAD3749-1GEA 0 CNY
TCIAD3749-1G TCIAD3749-5G
1,4-二氯酞嗪 ≥98.0% (by GC)
1,4-二氯酞嗪
分子式: C₈H₄Cl₂N₂
MDL编号: MFCD00006909
CAS号: 4752-10-7
EINECS: 225-275-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec37d4c18990d2d7494d9dbf277d8aaad787e993&vid=ec37d4c18990d2d7494d9dbf277d8aaad787e993&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR