N-Cyclohexyl-p-toluenesulphonamide ≥99.0% (by HPLC, total nitrogen)

供应商: TCI

C2032-25G C2032-500G
TCIAC2032-25GEA 0 CNY
TCIAC2032-25G TCIAC2032-500G
N-Cyclohexyl-p-toluenesulphonamide ≥99.0% (by HPLC, total nitrogen)
N-Cyclohexyl-p-toluenesulphonamide
分子式: C₁₃H₁₉NO₂S
分子量: 253.3605 g/mol
MDL编号: MFCD00014285
CAS号: 80-30-8
EINECS: 201-268-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29ce7466d9c78f191b52bdd56babe69f0a3aa61c&vid=29ce7466d9c78f191b52bdd56babe69f0a3aa61c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR