对甲苯磺酰甲基异腈 ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Danger

Synonyms: N-对甲苯磺酰甘氨酸, 对甲基苯磺酰甲基异腈, TosMIC

T1046-25G T1046-5G
TCIAT1046-25GEA 0 CNY
TCIAT1046-25G TCIAT1046-5G
对甲苯磺酰甲基异腈 ≥98.0% (by HPLC, total nitrogen)
对甲苯磺酰甲基异腈
分子式: C₉H₉NO₂S
分子量: 195.24 g/mol
MDL编号: MFCD00000005
CAS号: 36635-61-7
EINECS: 253-140-1
UN号: 2811
ADR: 6.1 III
Merck Index: 14,09556
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0007398119e07782b311d52fad0f6fb77065467a&vid=0007398119e07782b311d52fad0f6fb77065467a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR