Terephthalic acid dihydrazide ≥90.0% (by HPLC)
供应商: TCI
|
Warning
|
Synonyms:
Terephthalic Dihydrazide
T0758-25G
T0758-500G
TCIAT0758-25GEA
0
CNY
TCIAT0758-25G
TCIAT0758-500G
Terephthalic acid dihydrazide ≥90.0% (by HPLC)
Terephthalic acid dihydrazide
分子式:
C₈H₁₀N₄O₂ 分子量: 194.19 g/mol |
MDL编号:
MFCD00014762 CAS号: 136-64-1 EINECS: 205-253-2 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 90.0 area% |
Elemental analysis (Nitrogen) | 28.00 to 29.20 % |
Solubility in dilute HCl | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...