7-(Dimethylamino)-4-methylcoumarin ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

D3355-25G D3355-5G
TCIAD3355-25GEA 0 CNY
TCIAD3355-25G TCIAD3355-5G
7-(Dimethylamino)-4-methylcoumarin ≥98.0% (by titrimetric analysis)
7-(Dimethylamino)-4-methylcoumarin
分子式: C₁₂H₁₃NO₂
MDL编号: MFCD00006862
CAS号: 87-01-4
EINECS: 201-717-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=882e2cc9afcbff3e12eb914f3da3828a7a957258&vid=882e2cc9afcbff3e12eb914f3da3828a7a957258&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR