Methyl-4,4,4-trifluoroacetoacetate ≥96.0% (通过GC)
供应商: TCI
|
Warning
|
T1245-25G
T1245-5G
TCIAT1245-25GEA
0
CNY
TCIAT1245-25G
TCIAT1245-5G
Methyl-4,4,4-trifluoroacetoacetate ≥96.0% (通过GC)
Methyl-4,4,4-trifluoroacetoacetate
分子式:
C₅H₅F₃O₃ 沸点: 116°C 闪点: 40°C |
MDL编号:
MFCD00041004 CAS号: 83643-84-9 UN号: 3272 ADR: 3,III |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...