3,4′-二氨基二苯基甲烷 ≥98.0% (通过GC,滴定分析)

供应商: TCI
Warning

D1684-1G D1684-25G D1684-5G
TCIAD1684-1GEA 0 CNY
TCIAD1684-1G TCIAD1684-25G TCIAD1684-5G
3,4′-二氨基二苯基甲烷 ≥98.0% (通过GC,滴定分析)
3,4′-二氨基二苯基甲烷
分子式: C₁₃H₁₄N₂
MDL编号: MFCD00014778
CAS号: 19430-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62dcdb0b698880f6d012906fa832db8f5ad13ba6&vid=62dcdb0b698880f6d012906fa832db8f5ad13ba6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 84.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR